Learn More
Restek™ Super-Clean Gas Trapping System for LC/MS
Product Code. 13121452
Encompass_Preferred
Change view
Click to view available options
Type:
Gas Trapping System
Hydrocarbon Filter
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
13121452 Hydrocarbon Filter -
10111427 Gas Trapping System Each
13131452 Gas Trapping System -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13121452 Supplier Restek™ Supplier No. 22061
Quick-change cartridge system removing hydrocarbons efficiently from nitrogen
  • Changing filters is quick and easy
  • Up to 20L of hydrocarbon-free nitrogen per minute
  • Filters connected in parallel to handle high flows of LC/MS
Specifications
Type Hydrocarbon Filter
For Use With (Equipment) Super-Clean Gas Trapping System