missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Sulfinert Sample Loops

Product Code. 13137272
Encompass_Preferred
Click to view available options
Volume (Metric):
5 μL
10 μL
20 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2 mL
5 mL
This item is not returnable. View return policy

Product Code. 13137272

Brand: Restek™ 22850

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for samples containing low concentrations of sulfur or other active compounds. Sulfinert treatment of Restek Gas Sample Loops eliminate active sites in the valve or loop for better recovery of active compounds.

  • Sample loop sizes from 5μL to 5mL
  • 0.0625 in. fittings
  • 1mm I.D.
  • For “W Type” Valves

Specifications

Material Sulfinert treated
Volume (Metric) 5 mL
Type Gas Sample Loop
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.