Learn More
Restek™ Sulfinert Gas Sampling Valves
Product Code. 13021820
Encompass_Preferred
Click to view available options
Size:
10-Port
4-Port
6-Port
Product Code. 13021820
Brand: Restek™ 20585
Sulfinert-treated valves are ideal for samples containing low concentrations of sulfur or other active compounds.
  • “W Type” Valve
  • 0.0625 in. fittings
  • 0.40mm Port diameter
Specifications
For Use With (Equipment) 1/16 in. Fittings
Material Sulfinert-coated
Size 6-Port
Type Manual Sample Valve