missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Stx-CLPesticides Column Kits
Click to view available options
Diameter (Metric) Inner:
0.25 mm
Description
Includes
Stx-CLPesticides column, Stx-CLPesticides2 column, Universal Angled Y; Press-Tight connector and a Siltek-deactivated guard column
Requires
- Concentration in Solvent: 0.100g/dL each in water
- Volume: 1mL/ampule
- Shelf Life: 6 months
Compounds
- Acetaldehyde (75-07-0)
- Acetone (67-64-1)
- Acetonitrile (75-05-8)
- Ethanol (64-17-5)
- Ethyl acetate (141-78-6)
- 2-Propanol (Isopropanol) (67-63-0)
- Methanol (67-56-1)
- Methyl ethyl ketone (78-93-3)
- Baseline separations in less than 10 minutes
- Siltek surface deactivation enhances responses for endrin, DDT, methoxychlor
- Highly inert
- Stable to 330°C
Note:
Columns are not preconnected in this kit.
Order Info
Custom lengths and film thicknesses available.
Specifications
Specifications
| Max. Temperature | 330°C |
| Diameter (Metric) Inner | 0.25 mm |
| Stationary Phase | Stx™ -CLPesticides |
| For Use With (Application) | Organochlorine Pesticides Analysis, Herbicides Analysis |
| Length (Metric) | 30 m |
| Packing Material | Stx™ -CLPesticides |
| Includes | Rtx™-CLPesticides column, Rtx™-CLPesticides2 column, deactivated universal angled “Y” Press-Tight™ connector, guard column |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction