Learn More
Restek™ SSI TC-20 Tube Cutting Machine
Product Code. 13159722
Encompass_Preferred
Click to view available options
Type:
Replacement Cutting Wheel
SSI TC-20 Tube Cutting Machine
Unit Size:
Each
Product Code. 13159722
Brand: Restek™ 23030
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
This is the Replacement Cutting Wheel only.
Cuts 0.0625, 0.125, 0.25 in. tubing with inside diameter as small as 0.008 in.
  • Replacement Cutting Wheel
  • For use with the Restek SSI TC-20 Tube Cutting Machine
Specifications
Type Replacement Cutting Wheel
For Use With Restek SSI TC-20 Tubes Cutting Machine