Learn More
Restek™ Split/Splitless Injection Port
Description
- Manufactured from high-quality stainless steel
- Siltek treatment passivates the metal surface to ensure an inert pathway for the sample
Specifications
Specifications
| Quantity | Each |
| Product Type | Split/Splitless Injection Port |
| For Use With (Equipment) | Agilent 6890/6850 GCs, Large Canister Type Filter |
| Liner Material | Stainless Steel |