missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Split Liners for PerkinElmer Auto SYS GCs

Product Code. 13191652
Encompass_Preferred
Click to view available options
Deactivation:
Base Deactivated
Glass Wool
Intermediate-Polarity
Siltek
Geometry:
Cup
Cyclo
Laminar Cup
Precision
Straight
Product Type:
Inlet Liner
Split Inlet Liner
Split Liner
Quantity:
25/Pk.
5/Pk.
Each
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 13191652

Brand: Restek™ 20828

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For PerkinElmer Auto SYS and Clarus GCs equipped with split/splitless inlets.

  • Material: Borosilicate Glass
  • I.D.: 4mm
  • OD: 6.2mm
  • Length: 92.1mm

Specifications

Deactivation Intermediate-Polarity
For Use With (Application) For High MW Compounds
Geometry Laminar Cup
Quantity 5/Pk.
Length (Metric) 92.1 mm
Product Type Split Inlet Liner
Diameter (Metric) Inner 4 mm
Diameter (Metric) Outer 6.2 mm
For Use With (Equipment) For Perkin Elmer Auto SYS GCs
Liner Material Borosilicate Glass
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.