missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Split Liners for PerkinElmer Auto SYS GCs
Click to view available options
Deactivation:
Base Deactivated
Glass Wool
Intermediate-Polarity
Siltek
Geometry:
Cup
Cyclo
Laminar Cup
Precision
Straight
Product Type:
Inlet Liner
Split Inlet Liner
Split Liner
Quantity:
25/Pk.
5/Pk.
Each
Unit Size:
Pack of 5
Description
- Material: Borosilicate Glass
- I.D.: 4mm
- OD: 6.2mm
- Length: 92.1mm
Specifications
Specifications
| Deactivation | Intermediate-Polarity |
| For Use With (Application) | For High MW Compounds |
| Geometry | Cup |
| Quantity | 5/Pk. |
| Length (Metric) | 92.1 mm |
| Product Type | Split Inlet Liner |
| Diameter (Metric) Inner | 4 mm |
| Diameter (Metric) Outer | 6.2 mm |
| For Use With (Equipment) | For Perkin Elmer Auto SYS GCs |
| Liner Material | Borosilicate Glass |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction