missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ SPI Liners for Varian, Without Buffer
Click to view available options
Diameter (Metric) Inner:
0.53 mm
0.80 mm
Quantity:
25/Pk.
5/Pk.
Each
Deactivation:
Intermediate Polarity (IP)
Siltek
Unit Size:
Pack of 5
Description
- Geometry: Straight
- Material: Borosilicate Glass
- OD: 4.6mm
- Length: 54mm
Specifications
Specifications
| Deactivation | Intermediate Polarity (IP) |
| Geometry | Straight |
| Quantity | Each |
| Length (Metric) | 54 mm |
| Product Type | SPI Straight Inlet Liner |
| Diameter (Metric) Inner | 0.53 mm |
| Diameter (Metric) Outer | 4.6 mm |
| For Use With (Equipment) | Bruker/Varian GCs Equipped with SPI Inlets |
| Liner Material | Borosilicate Glass |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction