missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ SPI Liners for Varian, With Buffer
Click to view available options
Quantity:
25/Pk.
5/Pk.
Each
Deactivation:
Intermediate Polarity (IP)
Wool
Description
- Geometry: SPI with Buffer
- Material: Borosilicate Glass
- Deactivation: Intermediate Polarity (IP)
- Liner ID: 2.4mm
- Length: 54mm
- OD: 4.6mm
Specifications
Specifications
| Deactivation | Intermediate Polarity (IP) |
| Geometry | SPI with Buffer |
| Includes | Buffer |
| Quantity | 5/Pk. |
| Length (Metric) | 54 mm |
| Product Type | SPI Liner |
| Diameter (Metric) Inner | 2.4 mm |
| Diameter (Metric) Outer | 4.6 mm |
| For Use With (Equipment) | Bruker/Varian GCs Equipped with SPI Inlets |
| Liner Material | Borosilicate Glass |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction