Learn More
Restek™ Viton Replacement O-Rings for Septum-Purged Packed Column Port Weldment
Click to view available options
Packaging Quantity:
10/Pk.
Description
- Quantity: 10/Pk.
Specifications
Specifications
| Type | Viton O-Ring |
| Packaging Quantity | 10/Pk. |
| For Use With | Agilent 5890 GCs, Septum Purged Packed Column Port Weldment of Agilent 5890 GCs |