missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Shortix Capillary GC Column Cutter

Product Code. 13127402
Encompass_Preferred
Change view
Click to view available options
Type:
Capillary GC Column Cutter
Maintenance Kit
For Use With (Equipment):
0.25 to 0.53mm I.D. Tubing
Shortix Capillary GC Column Cutter
Unit Size:
Each
Product Code. Type For Use With (Equipment) unitSize
13127402 Capillary GC Column Cutter 0.25 to 0.53mm I.D. Tubing -
10152832 Capillary GC Column Cutter 0.25 to 0.53mm I.D. Tubing Each
10580613 Maintenance Kit Shortix Capillary GC Column Cutter -
13139722 Maintenance Kit Shortix Capillary GC Column Cutter -
4 options
This item is not returnable. View return policy

Product Code. 13127402

Brand: Restek™ 23026

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Consistently make precise, clean and square cuts with a diamond blade

  • Built-in magnifier to verify square cut
  • Use with 0.25 mm ID to 0.53 mm ID tubing (0.78 mm OD maximum)

Specifications

Type Capillary GC Column Cutter
Includes Built in magnifier
For Use With (Application) Gas Chromatography
For Use With (Equipment) 0.25 to 0.53mm I.D. Tubing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.