Learn More
Restek™ Septum Puller
Product Code. 13011570
Encompass_Preferred
Click to view available options
:
Septum puller for septa, o-rings, ferrules or fragments
Product Code. 13011570
Brand: Restek™ 20117
Only null left
Add to basket
Add to basket
Features one hooked end and one pointed end
  • Hooked end for removing septa and O-rings
  • Pointed end works well for removing stuck ferrules or debris
Specifications
Type Puller