Learn More
Restek™ Scoring Wafer with Handle
Product Code. 13167392
Encompass_Preferred
Click to view available options
:
Scoring Wafer with Ergonomic Rubber Handle and Ceramic Two-Sided Wafer
Product Code. 13167392
Brand: Restek™ 23015
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Straight edged for cutting fused silica tubing cleanly
  • Cuts both fused silica and metal tubing
  • Two-sided ceramic wafer
  • Serrated on one side and straight edged on the other
  • Ergonomic rubber handle
Specifications
Type Scoring Wafer