missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Rxi™-XLB GC Columns
Click to view available options
Length (Metric):
15 m
20 m
30 m
60 m
Diameter (Metric) Inner:
0.18 mm
0.25 mm
0.53 mm
Film Thickness:
0.1 μm
0.18 μm
0.25 μm
0.5 μm
1 μm
Description
- Especially well-suited for analysis of active, high molecular weight compounds with sensitive GC/MS systems including ion trap detectors
- Temperature range 30° to 360°C for 15m and 30m lengths: longer columns may have a slightly reduced maximum temperature
- Well-suited for U.S. EPA Method 525 for analysis of semivolatile compounds in drinking water
Pesticides, PCB cogeners
Specifications
Specifications
| Diameter (Metric) Inner | 0.25 mm |
| Stationary Phase | Rxi™-XLB |
| For Use With (Application) | Pesticide Analysis, PCB Congener Analysis, PAHs Analysis |
| Particle Size | 1 μm |
| Temperature Range | 30°C to 340/360°C |
| Film Thickness | 1 μm |
| Length (Metric) | 30 m |
| Polarity | Low polar |
| Coating | Fused Silica |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction