missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Rxi™-XLB GC Columns

Product Code. 12982499
Encompass_Preferred
Click to view available options
Length (Metric):
15 m
20 m
30 m
60 m
Diameter (Metric) Inner:
0.18 mm
0.25 mm
0.53 mm
Film Thickness:
0.1 μm
0.18 μm
0.25 μm
0.5 μm
1 μm
This item is not returnable. View return policy

Product Code. 12982499

Brand: Restek™ 13723

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

General-purpose columns exhibiting extremely low bleed

  • Especially well-suited for analysis of active, high molecular weight compounds with sensitive GC/MS systems including ion trap detectors
  • Temperature range 30° to 360°C for 15m and 30m lengths: longer columns may have a slightly reduced maximum temperature
  • Well-suited for U.S. EPA Method 525 for analysis of semivolatile compounds in drinking water

Pesticides, PCB cogeners

Specifications

Diameter (Metric) Inner 0.25 mm
Stationary Phase Rxi™-XLB
For Use With (Application) Pesticide Analysis, PCB Congener Analysis, PAHs Analysis
Particle Size 0.25 μm
Temperature Range 30°C to 340/360°C
Film Thickness 0.25 μm
Length (Metric) 30 m
Polarity Low polar
Coating Fused Silica
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.