Learn More
Restek™ RTX™-OPP Capillary Columns
Description
Using sophisticated computer modeling software, Restek created two stationary phases for separating the 53 organophosphorus pesticides (OPP) listed in EPA Method 8141. The extended upper temperature limit of these phases (330°C) allows analysts to bake out high molecular weight contamination typically associated with pesticide samples. The low-bleed columns are a perfect match for sensitive detection systems.
- Proprietary Crossbond phases
- Fused silica
- Low bleed makes it ideal for GC-FPD, GC-NPD, or GC-MS analyses
- Improved separation and reduced analysis time
- Stable to 330° to 340°C
Order Info
Custom lengths and film thicknesses available.
Specifications
Specifications
| Max. Temperature | 350°C |
| Diameter (Metric) Inner | 0.25 mm |
| Stationary Phase | Rtx™-OPP |
| For Use With (Application) | Organochlorine Pesticides Analysis, GC/NPD or GC/MS, Organochlorine Pesticides, Herbicides Analysis |
| Particle Size | 0.25 μm |
| Film Thickness | 0.25 μm |
| Length (Metric) | 30 m |
| Material | Stainless Steel |
| Coating | Fused Silica |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.