Learn More
Restek™ Ridgid Tubing Cutter
Product Code. p-3217667
Encompass_Preferred
Click to view available options
Type:
Replacement Cutting Wheels
Tubing Cutter, 1/8 in. or 1/4 in.
Packaging Quantity:
1/Pk.
2/Pk.
Product Code. 10583314
Brand: Restek™ 23011
Compact size for tight spaces
  • Excellent for cutting 1/8 in. or 1/4 in. metal tubing
  • Replaceable cutting wheel
Specifications
Type Tubing Cutter, 1/8 in. or 1/4 in.
For Use With (Application) Cutting 1/8 in. or 1/4 in. Tubing
Packaging Quantity 1/Pk.