Learn More
Restek™ Ridgid Tubing Cutter
Click to view available options
Type:
Replacement Cutting Wheels
Tubing Cutter, 1/8 in. or 1/4 in.
Packaging Quantity:
1/Pk.
2/Pk.
Description
- Excellent for cutting 1/8 in. or 1/4 in. metal tubing
- Replaceable cutting wheel
Specifications
Specifications
| Type | Replacement Cutting Wheels |
| For Use With (Application) | Cutting 1/8 in. or 1/4 in. Tubing |
| Packaging Quantity | 2/Pk. |