Learn More
Restek™ Ridgid Tubing Cutter
Description
- Excellent for cutting 1/8 in. or 1/4 in. metal tubing
- Replaceable cutting wheel
Specifications
Specifications
| Type | Tubing Cutter, 1/8 in. or 1/4 in. |
| For Use With (Application) | Cutting 1/8 in. or 1/4 in. Tubing |
| Packaging Quantity | 1/Pk. |