Learn More
Restek™ Rethreading Tools
Product Code. p-3186952
Encompass_Preferred
Click to view available options
For Use With:
1/16 in. Compression Fittings
1/4 in. Compression Fittings
1/8 in. Compression Fittings
7/16 in. Varian Injection Ports
Unit Size:
Each
Product Code. 13177392
Brand: Restek™ 23016
Only null left
Add to basket
Add to basket
Repair worn or damaged threads
  • Multiple uses (fittings, injectors, detectors, etc.)
  • Built-in guide to prevent cross-threading
Specifications
Type Rethreading Tool
For Use With 1/16 in. Compression Fittings