Learn More
Restek™ Rethreading Tools
Click to view available options
For Use With:
1/16 in. Compression Fittings
1/4 in. Compression Fittings
1/8 in. Compression Fittings
7/16 in. Varian Injection Ports
Unit Size:
Each
Description
- Multiple uses (fittings, injectors, detectors, etc.)
- Built-in guide to prevent cross-threading
Specifications
Specifications
| Type | Rethreading Tool |
| For Use With | 1/16 in. Compression Fittings |