Learn More
Restek™ Replacement Split/Splitless Weldment
Product Code. 13107142
Encompass_Preferred
Change view
Click to view available options
Material:
Stainless Steel
Product Code. Material
13107142 Stainless Steel
1 options
Product Code. 13107142 Supplier Restek™ Supplier No. 22686
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Direct replacement for Agilent 5890 and 6890/6850 GCs
  • Siltek™ treatment passivates the metal surface ensuring an inert pathway for the sample, delivering increased performance
  • High-quality stainless steel injector
  • Meets or exceeds Agilent original equipment performance
Specifications
For Use With (Equipment) Large Canister Type Filter, Agilent 6890/6850 Gcs
Material Stainless Steel
Product Type Split/Splitless Weldment