Learn More
Restek™ Replacement Split/Splitless Injection Ports: Replacement Weldments
Product Code. 13167132
Encompass_Preferred
Change view
Click to view available options
Product Type:
Shell Weldmen
Shell Weldment
Weldment
Weldment with EPC
Material:
Siltek Treated Stainless Steel
Stainless Steel
For Use With (Equipment):
Agilent 5890 GCs
Agilent 5890 GCs, Agilent 6890/6850 GCs
Agilent 6890/6850 GCs
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Material For Use With (Equipment)
13167132 Weldment with EPC Stainless Steel Agilent 6890/6850 GCs
13001680 Weldment Stainless Steel Agilent 5890 GCs, Agilent 6890/6850 GCs
13011680 Shell Weldment Stainless Steel Agilent 5890 GCs
13021680 Weldment Siltek Treated Stainless Steel Agilent 5890 GCs, Agilent 6890/6850 GCs
13031680 Shell Weldment Siltek Treated Stainless Steel Agilent 5890 GCs
13157132 Shell Weldmen Stainless Steel Agilent 6890/6850 GCs
13137132 Shell Weldment Siltek Treated Stainless Steel Agilent 6890/6850 GCs
13147132 Weldment with EPC Siltek Treated Stainless Steel Agilent 6890/6850 GCs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 13167132 Supplier Restek™ Supplier No. 22674
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Replacement weldments for Agilent 5890 and 6890/6850 GCs
  • Siltek™ treatment passivates metal surface ensuring an inert pathway for the sample, delivering increased performance
  • High-quality stainless steel injector
  • Meets or exceeds Agilent original equipment performance
Specifications
For Use With (Application) Gas Chromatography
For Use With (Equipment) Agilent 6890/6850 GCs
Material Stainless Steel
Product Type Weldment with EPC