Learn More
Restek™ Replacement Split/Splitless Injection Ports: Replacement Weldments
Product Code. 13001680
Encompass_Preferred
Change view
Click to view available options
Product Type:
Shell Weldmen
Shell Weldment
Weldment
Weldment with EPC
Material:
Siltek Treated Stainless Steel
Stainless Steel
For Use With (Equipment):
Agilent 5890 GCs
Agilent 5890 GCs, Agilent 6890/6850 GCs
Agilent 6890/6850 GCs
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Material For Use With (Equipment)
13001680 Weldment Stainless Steel Agilent 5890 GCs, Agilent 6890/6850 GCs
13011680 Shell Weldment Stainless Steel Agilent 5890 GCs
13021680 Weldment Siltek Treated Stainless Steel Agilent 5890 GCs, Agilent 6890/6850 GCs
13031680 Shell Weldment Siltek Treated Stainless Steel Agilent 5890 GCs
13157132 Shell Weldmen Stainless Steel Agilent 6890/6850 GCs
13137132 Shell Weldment Siltek Treated Stainless Steel Agilent 6890/6850 GCs
13147132 Weldment with EPC Siltek Treated Stainless Steel Agilent 6890/6850 GCs
13167132 Weldment with EPC Stainless Steel Agilent 6890/6850 GCs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 13001680 Supplier Restek™ Supplier No. 20265
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Replacement weldments for Agilent 5890 and 6890/6850 GCs
  • Siltek™ treatment passivates metal surface ensuring an inert pathway for the sample, delivering increased performance
  • High-quality stainless steel injector
  • Meets or exceeds Agilent original equipment performance
Specifications
For Use With (Application) Gas Chromatography
For Use With (Equipment) Agilent 5890 GCs, Agilent 6890/6850 GCs
Material Stainless Steel
Product Type Weldment