missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Replacement Orifices

Product Code. 13161972
Encompass_Preferred
Click to view available options
Size (Metric):
0.003 cm
Material:
Siltek Treated
Flow Rate:
2 to 4 cc/min.
This item is not returnable. View return policy

Product Code. 13161972

Brand: Restek™ 24233

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use these orifices with a Veriflo 423XL flow controller to change the flow range for alternative sampling times.

  • Available in stainless-steel or Siltek-treated material

Specifications

Flow Rate 2 to 4 cc/min.
Material Siltek Treated
Size (English) 0.0012 in.
Size (Metric) 0.003 cm
Type Replacement Orifice
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.