missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Replacement Inlet Cross-Disks for Agilent 6890 GCs

Product Code. 13154362
Encompass_Preferred
Click to view available options
Material:
Gold-plated
Siltek
Packaging Quantity:
10 Pack
2 Pack
This item is not returnable. View return policy

Product Code. 13154362

Brand: Restek™ 21009

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for high-flow split applications >500mL/min. on Agilent 5890 GCs.

  • Siltek treatment provides enhanced inertness versus stainless steel
  • Include washers
  • I.D.: 1.2mm
  • Available gold-plated or Siltek treated

Specifications

Type Inlet Cross-Disks
Diameter (Metric) Inner 1.2 mm
Packaging Quantity 2 Pack
For Use With Agilent 6890 GCs
Material Gold-plated
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.