missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Replacement Frits for Passive Air Sampling Kits

Product Code. 13161922
Encompass_Preferred
Click to view available options
Material:
Siltek Treated
Stainless Steel
This item is not returnable. View return policy

Product Code. 13161922

Brand: Restek™ 24171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in critical orifice fitting.

  • Size: 2μm
  • Available in stainless-steel or Siltek-treated material
  • Includes washers

Specifications

Includes Washers
Material Siltek Treated
Size 2 μm
Type Replacement Frit Filter
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.