Learn More
Restek™ Replacement Combination Vacuum/Pressure Gauges
Product Code. 13179992
Encompass_Preferred
Click to view available options
Pressure Range:
30 in. to 15 psig
30 in. to 30 psig
30 in. to 60 psig
Product Code. 13179992
Brand: Restek™ 24104
2 in. vacuum/pressure gauges
  • 316 stainless steel
  • 1/8 in. NPT fitting
  • Center back mount
Specifications
For Use With (Equipment) For canisters
Material 316 Stainless Steel
Pressure Range 30 in. to 30 psig
Product Type Vacuum/Pressure Gauge