Learn More
Restek™ Replacement Combination Vacuum/Pressure Gauges
Click to view available options
Pressure Range:
30 in. to 15 psig
30 in. to 30 psig
30 in. to 60 psig
Description
- 316 stainless steel
- 1/8 in. NPT fitting
- Center back mount
Specifications
Specifications
| For Use With (Equipment) | For canisters |
| Material | 316 Stainless Steel |
| Pressure Range | 30 in. to 15 psig |
| Product Type | Vacuum/Pressure Gauge |