Learn More
Restek™ Pressure Gauge Kit for Thermal Gas Purifiers
Click to view available options
Type:
Gas Pressure Gauge Kit
Description
- Includes 1/8 in. tee and 0-100 psi (0-689k Pa) gauge
Specifications
Specifications
| Type | Gas Pressure Gauge Kit |
| For Use With (Application) | Use an in-line pressure gauge to indicate when a thermal gas purifier getter tube should be replace |
| For Use With (Equipment) | Thermal gas purifier |
| Includes | 0.12 in. Tee and 0 to 100psi gauge |