Learn More
Restek™ Polypropylene Vial Inserts
Specially designed for 2.0mL, 11mm crimp-top and 2.0mL, 9mm short-cap, screw-thread vials.
£24.20
Specifications
Material Polypropylene
Quantity 100/Pk.
Type Vial Insert
View More Specs
Products 1
Product Code Brand Volume (Metric) Type Style Price Quantity & Availability  
Product Code Brand Volume (Metric) Type Style Price Quantity & Availability  
10045924
Encompass_Preferred
View Documents Promotion Details
Restek™
24520
250 μL Vial Insert Big Mouth without Spring
£24.20
Each
View alternative products
Save Up to 
»
Estimated Shipment: 25-10-2024
to see stock.
Add to basket
Description
Description
  • Big mouth
  • Pack of 100
Specifications
Specifications
Polypropylene
100/Pk.
Vial Insert