Learn More
Restek™ Pocket Magnifier
Product Code. 13051570
Encompass_Preferred
Click to view available options
:
Pocket Magnifier
Product Code. 13051570
Brand: Restek™ 20124
Only null left
Add to basket
Add to basket
Small and easy to handle
10x magnification makes it easy to see the column end to verify a square cut.
Specifications
Type Pocket Magnifier