missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Parker Precision Metering Valves
Click to view available options
Material:
Nickel-plated Brass
Stainless Steel
Thread Size:
1/4 in.
1/8 in.
Description
- Available in stainless steel or nickel-plated brass materials
- Available in two sizes; 0.25 in. or 0.125 in.
Specifications
Specifications
| Type | Valve |
| Thread Size | 1/4 in. |
| Material | Stainless Steel |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction