Learn More
Restek™ Open-End Wrench Sets
Product Code. p-3186833
Encompass_Preferred
Click to view available options
For Use With:
Shimadzu 17 A, 2010, 2014 Capillary Installation Gauge
Tightening Chromatography Fittings
Product Code. 13041740
Brand: Restek™ 20387
Only null left
Add to basket
Add to basket
For tightening a wide variety of chromatography fittings
Model 20387 Set:
  • 1/4 in. x 5/16 in.
  • 3/8 in. x 7/16 in.
  • 7/16 in. x 1/2 in.
  • 1/2 in. x 9/16 in.
Model 22334 Set (For use with Shimadzu 17A, 2010 and 2014 Capillary Installation Gauge):
  • 1/4 in. x 5/16 in.
  • 10mm x 11mm (0.39 in. x 0.43 in.)
Specifications
Type Open End Wrench Set
For Use With Tightening Chromatography Fittings