Learn More
Restek™ MINICYL Regulator
Click to view available options
Includes:
1/4 in. Tube Fittings
1/8 in. Tube Fittings
Description
Includes
0 to 60 psig (0 to 414kPa) gauge
- Laboratory use for air-drying glassware, sparging or evaporating solutions, and controlling pneumatic valves
- Constructed of lightweight aluminum with an elastomer diaphragm
- Choice of 0.32 or 0.64cm (0.125 or .25 in.) tube fittings
Specifications
Specifications
| Type | Fitting |
| Includes | 1/4 in. Tube Fittings |
| For Use With | Restek MINICYL |