Learn More
Restek™ Mini Hand Drill Set
Product Code. 13041570
Encompass_Preferred
Click to view available options
:
0.4, 0.5, and 0.8mm hand drills
Product Code. 13041570
Brand: Restek™ 20122
Only null left
Add to basket
Add to basket
Drill ferrules to the proper I.D. in seconds
  • Includes three drills
  • For use with 0.25, 0.32 and 0.53mm I.D. (0.4, 0.5 and 0.8mm O.D.) capillary columns
Specifications
Type Drill