Learn More
Restek™ Maintenance Supplies for Capillary Columns
Click to view available options
For Use With:
Capillary Columns
Description
- An alternative way to install capillary columns to the correct insertion distance recommended by the instrument manufacturer
- Useful for many laboratory tasks
Specifications
Specifications
| For Use With (Application) | Capillary Column Installation |
| Description | Slide-Lock Tweezers and 15 cm Compact Steel Ruler |
| For Use With | Capillary Columns |
| Overall Length (Metric) | 15 cm |
| Material | Steel |