Learn More
Restek™ Luer-Lock Valve Adapter
Description
- Quantity: 2/Pk.
Specifications
Specifications
| Type | Adapter |
| For Use With (Application) | Connecting a Luer Lock Fitting to a Rheodyne or Valco Valve |
| Packaging Quantity | 2/Pk. |