Learn More
Restek™ Injector Wrench for Agilent 5890/6890/6850 GCs
Description
- Use to remove the septum nut and weldments during GC maintenance
- Use the smaller end to remove the septum nut
- Use the larger end to tighten the split/splitless weldment nut
- High-quality stainless steel construction
Specifications
Specifications
| Type | Inlet Wrench |
| Material | Stainless Steel |
| For Use With (Equipment) | Agilent 5890/6890/6850 GCs |
Product Title
Haben Sie Verbesserungsvorschläge?