Learn More
Restek™ Injection Port Repair Tool for Agilent Split/Splitless Injection Ports
Description
- Removes contaminants and achieves a better seal
- Cleans critical inlet seal areas
- Kit includes - handle and brush adapter, five medium sanding disks, five fine sanding disks, a 6.5mm bore brush, and a 7mm bore brush
- Replacement Sanding Disks includes five each of fine and medium disks
Specifications
Specifications
| Type | Injection Port Repair Tool |
| Packaging Quantity | Each |
| Includes | Handle and brush adapter, five medium sanding disks, five fine sanding disks, a 6.5 mm bore brush, and a 7 mm bore brush |
| For Use With | Agilent Split/Splitless Injection Ports |