Learn More
Restek™ HPLC Piston Seal Insertion Tool
Click to view available options
:
Seal Insertion; HPLC
Description
- Simple function
- Tool
Specifications
Specifications
| Type | LC Piston Seal Insertion Tool |
| For Use With (Application) | Used to Simplifying Pump Maintenance by Seal Insertion and Cannot Mar The Surrounding Metal Surface of The Pump Housing |