missing translation for 'onlineSavingsMsg'
Learn More

Restek™ High-capacity and ECD Vent Traps

Product Code. 13161412
Encompass_Preferred
Change view
Click to view available options
Type:
ECD Vent Trap
High Capacity Split Vent Trap
Packaging Quantity:
1 Each
5/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Packaging Quantity
13161412 ECD Vent Trap 1 Each
12912609 High Capacity Split Vent Trap 1 Each
12922609 High Capacity Split Vent Trap 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13161412 Supplier Restek™ Supplier No. 22017
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes connecting lines and mounting kit.

  • High-Capacity Split Vent Trap reduces the release of hazardous materials from the capillary split vent into the lab
  • ECD Vent Trap reduces the release of hazardous materials from the ECD vent into the lab
  • Fittings: 1/8 in.
  • Dimensions: 15.2 x 2.5cm (6 x 1 in.)

Specifications

Connection Type 0.12 in. Fitting
Type ECD Vent Trap
Includes Connecting lines and mounting kit
Packaging Quantity 1 Each
Length (English) 6 in.
Length (Metric) 15.2 cm
Diameter (English) Outer 1 in.
Diameter (Metric) Outer 2.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.