missing translation for 'onlineSavingsMsg'
Learn More

Restek™ High-capacity and ECD Vent Traps

Product Code. 12922609
Encompass_Preferred
Click to view available options
Type:
ECD Vent Trap
High Capacity Split Vent Trap
Packaging Quantity:
1 Each
5/Pk.
This item is not returnable. View return policy

Product Code. 12922609

Brand: Restek™ 20699

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes connecting lines and mounting kit.

  • High-Capacity Split Vent Trap reduces the release of hazardous materials from the capillary split vent into the lab
  • ECD Vent Trap reduces the release of hazardous materials from the ECD vent into the lab
  • Fittings: 1/8 in.
  • Dimensions: 15.2 x 2.5cm (6 x 1 in.)

Specifications

Connection Type 0.12 in. Fitting
Type High Capacity Split Vent Trap
Includes Connecting lines and mounting kit
Packaging Quantity 5/Pk.
Length (English) 6 in.
Length (Metric) 15.2 cm
Diameter (English) Outer 1 in.
Diameter (Metric) Outer 2.5 cm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.