missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Glass Vial Inserts

Product Code. 15152875
Encompass_Preferred
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
350 μL
Style:
Bottom Spring
Flat Bottom
Glass Flange (step)
For Use With (Equipment):
Big Mouth Step Design Vials
Standard Mouth Vials
No. per Pack:
100
1000
This item is not returnable. View return policy

Product Code. 15152875

Brand: Restek™ 24513

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specially designed for 2 mL, 11 mm crimp top and 2 mL, 9 mm short cap, screw thread vials

  • Limited-volume inserts also available
  • Material: glass

Specifications

Material Glass
Volume (Metric) 50 μL
No. per Pack 100
Style Bottom Spring
For Use With (Equipment) Big Mouth Step Design Vials
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.