Learn More
Restek™ Gas Management System
Click to view available options
Type:
Gas Management System
Description
- Includes one each of moisture, hydrocarbon, and indicating oxygen trap
- Replacing traps is safe and easy
- Maximum flow: 150 cc/min. for optimal performance
- Includes fittings for 1/8 and 1/4 in. gas line
- Dimensions: 30.5 x 35.6 x 7.6cm
Specifications
Specifications
| For Use With (Application) | Removes moisture, hydrocarbons and oxygen from carrier gas |
| Includes | Moisture, hydrocarbon and indicating oxygen trap |
| Type | Gas Management System |