missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Fittings: Front Ferrules

Product Code. 13189692
Encompass_Preferred
Change view
Click to view available options
Material:
Brass
Stainless Steel
Size (Metric):
0.15 cm
0.31 cm
0.63 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Size (Metric)
13189692 Brass 0.15 cm
13181352 Stainless Steel 0.15 cm
13191352 Stainless Steel 0.31 cm
13101362 Stainless Steel 0.63 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13189692 Supplier Restek™ Supplier No. 21803

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.

  • Available in brass or stainless-steel materials

Specifications

Type Front Ferrule
Size (English) 0.062 in.
Size (Metric) 0.15 cm
For Use With (Equipment) Parker Fitting
Material Brass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.