missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Filter and Baseplate Kits
Single-position mounting baseplate with 0.125 in. inlet/outlet fittings (06-711-828)
£601.37
Product Code | Brand | Type | For Use With (Application) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Application) | Includes | Price | Quantity & Availability | |||
10169171
|
Restek™
22019 |
Carrier Gas Cleaning Kit | Filtering oxygen, moisture, and hydrocarbon impurities | Mounting base plate, 0.12 in. inlet/outlet fittings, and oxygen/moisture/hydrocarbon triple filter |
£601.37
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13171412
|
Restek™
22019 |
Carrier Gas Cleaning Kit | Filtering oxygen, moisture, and hydrocarbon impurities | Mounting base plate, 0.12 in. inlet/outlet fittings, and oxygen/moisture/hydrocarbon triple filter |
£601.37
Each |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13191412
|
Restek™
22021 |
Fuel Gas Purification Kit | Filtering for moisture and hydrocarbon impurities | Mounting base plate, 0.12 in. inlet/outlet fittings, and hydrocarbon/moisture filter |
£601.37
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Carrier Gas Cleaning Kit
- Single-position mounting baseplate with 0.125 in. inlet/outlet fittings (06-711-828)
- Triple filter for oxygen, moisture, and hydrocarbon impurities (06-711-824)
- Single-position mounting baseplate with 0.125 in. inlet/outlet fittings (06-711-828)
- Fuel gas filter for moisture and hydrocarbon impurities (06-711-826)
Specifications
Filter and Baseplate Kit |