Learn More
Restek™ FID/Injector Cleaning Kit
Product Code. 10793222
Encompass_Preferred
Click to view available options
:
Cleaning Tool Kit
Product Code. 10793222
Brand: Restek™ 20120
Only null left
Add to basket
Add to basket
Includes
  • Nylon tube brushes (1/8 in., 3/16 in., 1/4 in.)
  • Pipe cleaner
  • Stainless steel tube brushes (3/8 in., 3/16 in., 1/4 in.)
  • Stainless steel surface brush
  • Stainless steel jet reamers
  • Emery cloth
Use with any injection port
  • Cleaning tool kit
Specifications
Type Tool Kit