Learn More
Restek™ Ferrule Removal Kit
Description
- The tapered tools in this kit have teeth designed to grip and remove fused silica adaptor ferrules that have become stuck in the fitting detail
Specifications
Specifications
| For Use With (Equipment) | 1/32 and 1/16 in. Adaptor Ferrules |